Bottles, narrow neck, round, PS, with dip tube, Corning®
Supplier: Corning
Certificates
About this item
Polystyrene, clear with HDPE screw cap.
- Easy aseptic transfer of liquid
- Good chemical resistance
- Good temperature resistance
Sterile, double-bagged and preassembled. The preassembled transfer caps have two ports. One port ends in a 0,2 µm filter. The other port is heat sealable flexible tubing, ends in a male MPC or male luer lock connection.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Catalog No:
- CORN11680
- CORN11665
- CORN11655
- CORN11681
- CORN11651
- CORN11666
- CORN11656
- CORN11650
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk