Order Entry
Export
Orders LinkContactUsLinkComponent
Heating tapes, HBV

Heating tapes, HBV

Supplier:   HORST
Heating tapes, HBV
Supplier:  HORST
Certificates
Heating tapes, HBV

7 Options Available Below

Product Details & Documents

The heating tape comes with 1,4301 (V2A) stainless steel protective braid and multiple electrical glass-yarn insulation. This braid minimises the release of fibres.

  • Nominal temperature: 450 °C
  • Nominal voltage: ~230 V
  • Thickness×Breadth: 5×30 mm
Product Family Options

Product Information

  • Availability
    Price
  • HORS020331
    Heating tape HBV, 0,5 m length, 100 W
    Heating tape HBV, 0,5 m length, 100 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • HORS020332
    Heating tape HBV, 1,0 m length, 250 W
    Heating tape HBV, 1,0 m length, 250 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 160,50 €
    160,50 €
    1 * 1 items
       160,50 €

  • HORS020333
    Heating tape HBV, 1,5 m length, 350 W
    Heating tape HBV, 1,5 m length, 350 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • HORS020334
    Heating tape HBV, 2,0 m length, 500 W
    Heating tape HBV, 2,0 m length, 500 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • HORS020335
    Heating tape HBV, 2,5 m length, 600 W
    Heating tape HBV, 2,5 m length, 600 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • HORS020336
    Heating tape HBV, 3,0 m length, 750 W
    Heating tape HBV, 3,0 m length, 750 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • HORS020337
    Heating tape HBV, 5,0 m length, 1250 W
    Heating tape HBV, 5,0 m length, 1250 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items