ContactUsLinkComponent
VWR®, Tubing Clamp Kit
  VWRU21732-000
undefined
VWR®, Tubing Clamp Kit
  VWRU21732-000
  • Description:
    Tubing clamp kit incl. 13 clamps of different sizes
  • Environmentally Preferable:
  • Pk:
    1
Tubing clamps for pressure, vacuum, and liquid applications when a tight seal is required. The chemical- and corrosion-resistant nylon clamps are ideal for use in a wide range of temperatures.
  • Works with all types of plastic and rubber tubing
  • May be chemically and gas sterilised for biomedical or food applications
  • Clamps securely fasten tubing on connectors, pumps, glassware, and filtration units
  • One-piece reusable clamp squeezes shut for a leakproof seal; to release the clamp, press down on side
 : Kit is supplied with 13 clamp sizes to accommodate tubing. Five of each size are supplied.
Clamp int.Ø. range for these 13 sizes: 0,246 to 0,290", 0,360 to 0,406", 0,410 to 0,468", 0,475 to 0,536", 0,538 to 0,608", 0,597 to 0,665", 0,670 to 0,780", 0,730 to 0,830", 0,870 to 0,975", 0,953 to 1,093", 1,031 to 1,187", 1,250 to 1,425", and 1,406 to 1,610".