Order Entry
Export
ContactUsLinkComponent
Regenerating ointment, ENZBORN® Zinc
Regenerating ointment, ENZBORN® Zinc
Catalog # EIMM62018
Supplier:  EIMERMACHER
Regenerating ointment, ENZBORN® Zinc
Catalog # EIMM62018
Supplier:  EIMERMACHER

Specifications

  • Description:
    Regenerating ointment, ENZBORN® Zinc, tube, 50 ml
  • Pk:
    50 mL

Specifications

About this item

ENZBORN® Zinc Ointment promotes the regeneration of the skin. It is produced according to a formula which has prooved its success for decades. The highly concentrated composition helps the skin to regenerate even better after minor wounds and skin abrasions.

  • Highly effective components regulate the moisture and care for the skin
  • Can be applied easily
  • dermatest® very good

Suitable for daily use.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904