Wipes, pre-saturated
229×229 mm, 30 wipes/bag
Catalog # EPTA6-LS7030-PC-24
Supplier: ESSENTRA POROUS TECHNOLOGIES
Jump to:
Product Details & Documents
These polyester/cellulose wipes are pre-saturated with 70% IPA and 30% DIW. They are ideal for general and maintenance applications, as well as for industrial cleaning.
- Pre-wetted with alcohol
- Resealable pouch for easy dispensing
- Excellent as a degreaser
- Made in the USA
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Packed:30 wipes/bag
- L×W:229×229 mm



