WetGrip labels
Supplier: DIVERSIFIED BIOTECH
Jump to:
About this item
These WetGrip labels grip to moist, frosty or frozen surfaces.
- Temperature resistant to −80 °C
- Compatible with centrifuge rotors
- Resistant to chemicals and solvents
- Adheres to plastics, glass, metals and even paperboard boxes
Available in white laser sheets, white direct thermal rolls, or white hand writable rolls.
9 Options Available Below
Product Details & Documents
These WetGrip labels grip to moist, frosty or frozen surfaces.
- Temperature resistant to −80 °C
- Compatible with centrifuge rotors
- Resistant to chemicals and solvents
- Adheres to plastics, glass, metals and even paperboard boxes
Available in white laser sheets, white direct thermal rolls, or white hand writable rolls.
Product Family Options
Direct thermal labels
Product Information
- MaterialPrinting methodColourDimensionsAvailabilityPrice
- DIVEGRDT-1000WetGrip printer labels for 0,5 ml tubesWetGrip printer labels for 0,5 ml tubesPaperDirect thermalWhite27×13 mmLog in for availability1 * 1.000 items128,00 €
- DIVEGRDT-2000WetGrip printer labels for 1,5 ml tubesWetGrip printer labels for 1,5 ml tubesPaperDirect thermalWhite38×13 mmLog in for availability1 * 750 items130,00 €
- DIVEGRDT-3000WetGrip printer labels for 2,0 ml tubesWetGrip printer labels for 2,0 ml tubesPaperDirect thermalWhite38×19 mmLog in for availability1 * 750 items119,00 €
- DIVEGRDT-6000WetGrip printer labels, with 9,5 mm spot for 1,5 ml tubesWetGrip printer labels, with 9,5 mm spot for 1,5 ml tubesPaperDirect thermalWhite38×13 mmLog in for availability1 * 500 items157,00 €
- DIVEGRDT-4000WetGrip printer labels for 0,5 ml tubesWetGrip printer labels for 0,5 ml tubesPaperDirect thermalWhite10 mmLog in for availability1 * 1.000 items176,00 €
Laser labels
Product Information
- MaterialPrinting methodColourDimensionsAvailabilityPrice
- DIVEGRLA-1000WetGrip printer labels for microscope slidesWetGrip printer labels for microscope slidesPaperLaserWhite22×22 mmLog in for availability1 * 2.400 items184,00 €
- DIVEGRLA-1050WetGrip printer labels for 1,5 - 2,0 ml tubesWetGrip printer labels for 1,5 - 2,0 ml tubesPaperLaserWhite38×19 mmLog in for availability1 * 1.500 items184,00 €
- DIVEGRLA-2016WetGrip printer labels for 1,5 ml tubesWetGrip printer labels for 1,5 ml tubesPaperLaserWhite33×13 mmLog in for availability1 * 1.700 items184,00 €
- DIVEGRLA-2380WetGrip printer labels for 0,5 ml tubesWetGrip printer labels for 0,5 ml tubesPaperLaserWhite24×13 mmLog in for availability1 * 2.380 items184,00 €