Orders LinkContactUsLinkComponent
Caps for screw neck vials
Caps for screw neck vials
Screw cap, Alliance systems, blue PTFE/white silicone, pre-slit
Catalog # WATE186000305
Supplier:   WATERS
Caps for screw neck vials
Catalog # WATE186000305
Supplier:  WATERS
Caps for screw neck vials
Screw cap, Alliance systems, blue PTFE/white silicone, pre-slit
Price
84,20 €
Log in for availability
1 * 100 items
Product Details & Documents
Dark blue PE screw cap with bonded slit septum for 12×32 mm vials.
Specifications
  • For:
    Alliance systems
  • Septum:
    blue PTFE/white silicone, pre-slit
  • Type:
    Screw cap