Chemical resistant overalls, Body Plus
BESF01073A010XL
- Clothing Size:XL
- Closure type:Zip closure
- Garment application:Laboratory,Industrial
- Colour:White
- Seq:01
- Model:Body Plus
- Pk:40
- Weight:50 g/m²
These hooded overalls are made of breathable SMS fabric. They offer protection against hazardous dust and light liquid chemical splashes and sprays.
- Elastic wrists, ankles and back Moisture wicking properties Zio closure with adhesive flap cover
Colour: White
Weight: 50 g/m²
: Type 5/6; Cat. III