Safety boots, Wellingtons
Supplier: M S A
Certificates
About this item
These anti-static, water resistant protective boots have a profiled outer sole (oil and petrol resistant).
- Nail-puncture resistant midsole made from steel
- Steel toecaps (200 J)
Suitable for use on construction sites.
Colour: Yellow
Certifications: EN ISO 20345; S5