Order Entry
Export
ContactUsLinkComponent
 
Freezer and Food Storage Container
  216-3429
undefined
Freezer and Food Storage Container
  216-3429

 

  • Capacity:
    5,6 l
  • Description:
    Set of 3 boxes
  • W×D×H:
    295×195×75 mm
  • Pk:
    1

 

 

Made of plastic.

  • Food-safe and dishwasher-safe Temperature: -40 °C to + 90 °C Jars transparent, colourful lid

Multipurpose boxes for a wide range of storage and transport applications.