Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Swift™
Swift™ membrane stain
Swift™ membrane stain
Health
Swift™ membrane stain
Catalog # 786-677
Supplier:
G-Biosciences
CAS Number:
Expand image
Health
Swift™ membrane stain
Catalog # 786-677
Supplier:
G-Biosciences
CAS Number:
Specifications
Pk:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
Documentation
:
:
GHS08
Product Details
Documentation