Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Transfer Tubing
Tubing, PVC, coiled
Tubing, PVC, coiled
Tubing, PVC, coiled
Supplier:
LEYBOLD
Certificates
Expand image
Tubing, PVC, coiled
Supplier:
LEYBOLD
About this item
PVC, clear.
Without flanges
Good chemical resistance
Good temperature resistance
PVC coiled vacuum tubing.