Rotary vane vacuum pumps, TRIVAC B
Supplier: LEYBOLD
Certificates
About this item
These TRIVAC B two stage rotary vane vacuum pumps has wide range of applications. The inner body is assembled from individual parts without sealing components. The parts are pinned to ensure easy disassembly and reassembly of the parts.
- Highly leak tight and water vapor tolerance
- Built-in oil pump, pressure lubricated sliding bearings
- Vertical or horizontal intake and exhaust ports
- Anti-suck back valve controlled via the oil pressure
- Single phase motor
All controls and oil sight glass are located on the front face, they are ideal as backing pump for medium and high vacuum applications, because of low oil back streaming.
Certifications: Certified in accordance with 94/9/EG (ATEX).
Delivery: Supplied with small flanges, carrying handle, oil, mains cord and plug.