Terry tool tubing cutters
Catalog # HYPE60182-510
Supplier: Thermo Fisher Scientific
Specifications
- Description:Terry tool tubing cutter, for 1/8" stainless steel tubing
- Pk:1
Specifications
About this item
Tubing cutter cuts ¹/₈" stainless steel tubing.
Produce clean, 90° cuts of ¹/₈" stainless steel tubing.