Orders LinkContactUsLinkComponent
Heating tapes, HBS
Supplier:   HORST
Heating tapes, HBS
Supplier:  HORST
Certificates
Heating tapes, HBS
8 Options Available Below
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Product Family Options
Product Information
  • Availability
    Price
  • HORS020302
    Heating tape HBS, 1 m length, 250 W
    Heating tape HBS, 1 m length, 250 W
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m
  • HORS020303
    Heating tape HBS, 1,5 m length, 350 W
    Heating tape HBS, 1,5 m length, 350 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • HORS020304
    Heating tape HBS, 2 m length, 500 W
    Heating tape HBS, 2 m length, 500 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 159,50 €
    159,50 €
    1 * 1 items
       159,50 €
  • HORS020305
    Heating tape HBS, 2,5 m length, 600 W
    Heating tape HBS, 2,5 m length, 600 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • HORS020306
    Heating tape HBS, 3 m length, 750 W
    Heating tape HBS, 3 m length, 750 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 216,00 €
    216,00 €
    1 * 1 items
       216,00 €
  • HORS020311
    Heating tape HBS, 4 m length, 900 W
    Heating tape HBS, 4 m length, 900 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • HORS020307
    Heating tape HBS, 5 m length, 1250 W
    Heating tape HBS, 5 m length, 1250 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • HORS020308
    Heating tape HBS, 7 m length, 1550 W
    Heating tape HBS, 7 m length, 1550 W
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items