Screw caps, BOLA
Catalog # BOHLH988-40
Supplier: Bohlender
Specifications
- Thread:S 40
- Colour:Anthracite
- Pk:1
Specifications
About this item
Screw caps, made of PPS, with handy knurl, with integrated PTFE/silicone gasket for compensation of unevenness on the sealing surface.
- Universal chemical resistance
- Temperature resistance: from –20 to +230 °C
After assembly, the product is only exposed to PTFE. The cap has high mechanical and thermal (up to +230 °C) capacitites.