Drum stands, PE-FB
Supplier: LACONT UMWELTTECHNIK
Jump to:
About this item
PE, black or yellow.
- 1-drum models made of black PE
- 2-drum models made of yellow PE
For horizontal storage of steel drums (60 or 200 L capacity)
5 Options Available Below
Product Details & Documents
PE, black or yellow.
- 1-drum models made of black PE
- 2-drum models made of yellow PE
For horizontal storage of steel drums (60 or 200 L capacity)
Product Family Options
Product Information
- ColourW×D×HAvailabilityPrice
- LACOP70-7111-ADrum stands, Black, for horizontal storage of steel drums, FB 1/60Drum stands, Black, for horizontal storage of steel drums, FB 1/60Black390×515×390 mmLog in for availability1 * 1 items
- LACOP70-7112-ADrum stands, Black, for horizontal storage of steel drums, FB 1/200Drum stands, Black, for horizontal storage of steel drums, FB 1/200Black580×790×590 mmLog in for availability1 * 1 items
- LACOP70-7115-ADrum stands, Black, for horizontal storage of steel drums, FB 1-combiDrum stands, Black, for horizontal storage of steel drums, FB 1-combiBlack490×505×390 mmLog in for availability1 * 1 items
- LACOP70-7113-ADrum stands, Yellow, for horizontal storage of steel drums, FB 2/60Drum stands, Yellow, for horizontal storage of steel drums, FB 2/60Yellow860×385×315 mmLog in for availability1 * 1 items
- LACOP70-7114-ADrum stands, Yellow, for horizontal storage of steel drums, FB 2/200Drum stands, Yellow, for horizontal storage of steel drums, FB 2/200Yellow1245×585×390 mmLog in for availability1 * 1 items