Drum sumps, PE-100-EP
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
Rotationally-molded body of black PE.
- Circumferential stabilising ribs
- Easily removable PE grid support
Ideal for storing water-endangering and aggressive media.
Rotationally-molded body of black PE.
Ideal for storing water-endangering and aggressive media.