Plugs, PTFE, BOLA
Supplier: Bohlender
Certificates
About this item
Made of PTFE, white.
- Universal chemical resistance
- Temperature resistance: from –200 to +260 °C
For closing unused ports of miniature fittings.
Made of PTFE, white.
For closing unused ports of miniature fittings.