ContactUsLinkComponent
Packaging bags, MELAfol®
Packaging bags, MELAfol®
  MELA00751
undefined
Packaging bags, MELAfol®
  MELA00751
  • Description:
    MELAfol® packaging pouches with sterilization indicator
  • L×W:
    75×250 mm
  • Pk:
    1.000
MELAfol® is a combination packaging solution. It is sealed against all germs, tear resistant, hot sealable, crease-resistant and easy to open (with a 'peelable' seal seam).
  • It has a handling indicator in accordance with EN 868-5 which colours from rose to brown upon sterilisation
  • The packaging can be opened through pulling apart the side seal sealing
  • Storage length in accordance with DIN 58953 part 7: up to 6 months