0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
:
Thermo Fisher Scientific
n-Dodecyl-β-D-maltoside
:
Thermo Fisher Scientific
:
69227-93-6
:
:
C₂₄H₄₆O₁₁
:
MFCD00043012
:
510.6 g/mol
:
224…226 °C
:
GHS07
:
Freezer
Thermo Scientific Acroseal 25 Years