Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Actinomycin D
Actinomycin D ≥98%
Actinomycin D ≥98%
Actinomycin D ≥98%
Supplier:
Cayman Chemical
Certificates
Expand image
Actinomycin D ≥98%
Supplier:
Cayman Chemical
About this item
CAS Number:
50-76-0
:
Formula:
C₆₂H₈₆N₁₂O₁₆
MDL Number:
MFCD00005033
:
1255.5 g/mol
Storage Temperature:
Freezer
UN:
3462
ADR:
6.1,II