Ethynyltributyltin
Glass bottle for solids, 25 g
Catalog # COBBSS-0607-25G
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
Synonyms: Ethynyltri-n-butylstannane, Tributylethynylstannane
- CAS Number:994-89-8
- Formula:C₁₄H₂₈Sn
- MDL Number:MFCD00009420
- Molecular Weight:315.09 g/mol
- UN:2788
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



