Magnesium chloride
Catalog # ROTHKK36.2
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
About this item
:
:95.21 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:715 °C
Density:2.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106