Methyl blue
Glass bottle for solids, 10 g
Catalog # ROTH5750.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Aniline blue sodium salt, [[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2), Poirriers blue, Cotton blue, Water blue, C.I. 42780, Aniline blue, water soluble, Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonate, Acid Blue 93
- CAS Number:28983-56-4
- Formula:C₃₇H₂₇N₃Na₂O₉S₃
- MDL Number:MFCD00058509
- Molecular Weight:799.81 g/mol
- Merck Index:13,06055
- Storage Temperature:Ambient
- EINECS:249-352-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



