3,5-Dibromophenol ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
- CAS Number:626-41-5
- Formula:C₆H₄Br₂O
- MDL Number:MFCD06411376
- Molecular Weight:251.91 g/mol
- Boiling Pt:274 °C (1013 hPa)
- Melting Pt:79…83 °C
- Density:2.1 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:626-41-5
- Formula:C₆H₄Br₂O
- MDL Number:MFCD06411376
- Molecular Weight:251.91 g/mol
- Boiling Pt:274 °C (1013 hPa)
- Melting Pt:79…83 °C
- Density:2.1 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- UN:2811
- ADR:6.1,III



