Order Entry
Export
Orders LinkContactUsLinkComponent
Diallyl disulphide

Diallyl disulphide

Irritant
Supplier:   Cayman Chemical
Diallyl disulphide
Supplier:  Cayman Chemical
Certificates
Diallyl disulphide

3 Options Available Below

Product Details & Documents
Synonyms:  Diallyldisulphide, Allyl disulphide
  • CAS Number:2179-57-9
  • Formula:C₆H₁₀S₂
  • MDL Number:MFCD00008656
  • Molecular Weight:146.28 g/mol
  • Boiling Pt:198 °C (1013 hPa)
  • Density:1.0237 g/cm³ (15 °C)
  • Storage Temperature:Refrigerator
  • EINECS:218-548-6
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 34,00 €
    34,00 €
    1 * 100 mg
       34,00 €

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 81,00 €
    81,00 €
    1 * 500 mg
       81,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 126,00 €
    126,00 €
    1 * 1 g
       126,00 €