Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,5'-Dimethyl-2,2'-bipyridyl
5,5'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
5,5'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
5,5'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
5,5'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
1762-34-1
:
Formula:
C₁₂H₁₂N₂
MDL Number:
MFCD01740554
:
M-Clarity™ quality level = MQ100
:
184.24 g/mol