Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethylthiophene
3,4-Dimethylthiophene
3,4-Dimethylthiophene
3,4-Dimethylthiophene
Supplier:
Thermo Scientific
Certificates
Expand image
3,4-Dimethylthiophene
Supplier:
Thermo Scientific
About this item
CAS Number:
632-15-5
:
Formula:
C₆H₈S
MDL Number:
MFCD01764731
:
112.1900 g/mol