Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Di-o-tolylguanidine
1,3-Di-o-tolylguanidine, Sigma-Aldrich®
1,3-Di-o-tolylguanidine, Sigma-Aldrich®
1,3-Di-o-tolylguanidine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,3-Di-o-tolylguanidine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
97-39-2
:
Formula:
C₁₅H₁₇N₃
MDL Number:
MFCD00008513
:
239.32 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
202-577-6