Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-5-sulphamoylbenzoic acid
2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%
2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%
2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%
B24589.18
:
Thermo Fisher Scientific
:
2,4-Dichloro-5-sulphamoylbenzoic acid ≥98%
B24589.18
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
50 g
:
:
270.09 g/mol
:
230…232 °C (dec.)
:
220-358-3
:
GHS05
:
GHS07
:
Ambient
:
2736-23-4
:
C₇H₅Cl₂NO₄S
:
MFCD00007931