1,3-Diphenoxybenzene, Sigma-Aldrich®
Catalog # SIAL232637-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:262.31 g/mol
:M-Clarity™ quality level = MQ100
EINECS:222-181-7
CAS Number:3379-38-2
Formula:C₁₈H₁₄O₂
MDL Number:MFCD00003036