Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
15-Hydroxypentadecanoic acid
15-Hydroxypentadecanoic acid, Sigma-Aldrich®
15-Hydroxypentadecanoic acid, Sigma-Aldrich®
SIAL392979-5G
:
Merck
:
15-Hydroxypentadecanoic acid, Sigma-Aldrich®
SIAL392979-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
258.40 g/mol
:
M-Clarity™ quality level = MQ200
:
225-026-1
:
4617-33-8
:
C₁₅H₃₀O₃
:
MFCD00039535