Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
15-Hydroxypentadecanoic acid
15-Hydroxypentadecanoic acid ≥99%
15-Hydroxypentadecanoic acid ≥99%
15-Hydroxypentadecanoic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
15-Hydroxypentadecanoic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
4617-33-8
:
Formula:
C₁₅H₃₀O₃
MDL Number:
MFCD00039535
:
258.40 g/mol
Melting Pt:
84…88 °C
Storage Temperature:
Ambient
EINECS:
225-026-1
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB21670
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation