Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
11-Hydroxyundecanoic acid
11-Hydroxyundecanoic acid, Sigma-Aldrich®
11-Hydroxyundecanoic acid, Sigma-Aldrich®
11-Hydroxyundecanoic acid, Sigma-Aldrich®
SIAL524395-1G
:
Merck
:
11-Hydroxyundecanoic acid, Sigma-Aldrich®
SIAL524395-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
202.2900 g/mol
:
M-Clarity™ quality level = MQ100
:
222-932-9
:
3669-80-5
:
C₁₁H₂₂O₃
:
MFCD00041564