4,4'-Difluorobenzhydryl chloride, Sigma-Aldrich®
Catalog # SIAL360724-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:238.66 g/mol
:M-Clarity™ quality level = MQ100
EINECS:248-201-4
CAS Number:27064-94-4
Formula:C₁₃H₉ClF₂
MDL Number:MFCD00044329