
4-(Chloromethyl)-1,2-diphenylethane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:80676-35-3
:
Formula:C₁₅H₁₅Cl
MDL Number:MFCD02093918
:230.73 g/mol
Melting Pt:38…42 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05