2,6-Dichloro-3-cyano-4-picoline, Sigma-Aldrich®
Catalog # SIAL643521-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:187.03 g/mol
EINECS:212-873-7
CAS Number:875-35-4
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD00052633