2,6-Dichloro-3-cyano-4-picoline
Catalog # MOLEM39195370
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.03 g/mol
EINECS:212-873-7
CAS Number:875-35-4
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD00052633