Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
Supplier:
Thermo Scientific
Certificates
Expand image
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol
Supplier:
Thermo Scientific
About this item
CAS Number:
59108-13-3
:
Formula:
C₁₀H₄Cl₂F₃NO
MDL Number:
MFCD00173851
:
282.0500 g/mol