Order Entry
Export
ContactUsLinkComponent
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98%
Irritant
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98%
Catalog # B20259.09
CAS Number:  
Irritant
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene ≥98%
Catalog # B20259.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

CAS Number:1992-15-0
 :
Formula:C₁₂H₆F₁₂O₂
MDL Number:MFCD00042091
 :410.16 g/mol
Boiling Pt:100 °C (20 mmHg)
Melting Pt:92…95 °C
 :GHS07
Storage Temperature:Ambient