Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4'-Methylenebis(2-chloroaniline)
4,4'-Methylenebis(2-chloroaniline), Sigma-Aldrich®
4,4'-Methylenebis(2-chloroaniline), Sigma-Aldrich®
4,4'-Methylenebis(2-chloroaniline), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
4,4'-Methylenebis(2-chloroaniline), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
101-14-4
:
Formula:
C₁₃H₁₂Cl₂N₂
MDL Number:
MFCD00047829
:
M-Clarity™ quality level = MQ100
EINECS:
202-918-9