
Aniline-2,5-disulphonic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-44-2
:
Formula:C₆H₇NO₆S₂
MDL Number:MFCD00035753
:253.25 g/mol
Melting Pt:>300 °C
EINECS:202-669-6
:GHS07
Storage Temperature:Ambient