
4,6-Dichloro-2-(methylthio)pyrimidine ≥98%
Catalog # B21416.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:6299-25-8
:
Formula:C₅H₄Cl₂N₂S
MDL Number:MFCD00006086
:195.07 g/mol
Boiling Pt:135…136 °C (14 mmHg)
Melting Pt:38…42 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:228-577-6
ADR:8,II
:GHS05