2,5-Diaminohydroquinone dihydrochloride, Sigma-Aldrich®
Catalog # SIAL33040-1G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.06 g/mol
CAS Number:24171-03-7
Formula:C₆H₁₀Cl₂N₂O₂
MDL Number:MFCD00239416