1,1-Cyclohexanediacetic acid, Sigma-Aldrich®
Catalog # SIAL171344-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:200.23 g/mol
:M-Clarity™ quality level = MQ100
EINECS:224-427-9
CAS Number:4355-11-7
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00001519