5,7-Dichloro-2-methyl-8-quinolinol, Sigma-Aldrich®
Catalog # SIAL130532-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:228.0747 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-789-3
CAS Number:72-80-0
Formula:C₁₀H₇Cl₂NO
MDL Number:MFCD00023984