Order Entry
Export
ContactUsLinkComponent
 
Corrosive
Irritant
2,6-Dichloro-4-methylpyridine 95%
Catalog # H61767.03
CAS Number:  
Corrosive
Irritant
2,6-Dichloro-4-methylpyridine 95%
Catalog # H61767.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :162.02 g/mol
 :GHS05
 :GHS07
Storage Temperature:Freezer
CAS Number:39621-00-6
Formula:C₆H₅Cl₂N
MDL Number:MFCD09264302