Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-3-methylpyridine
2,6-Dichloro-3-methylpyridine ≥98%
2,6-Dichloro-3-methylpyridine ≥98%
H61271.03
:
Thermo Fisher Scientific
:
2,6-Dichloro-3-methylpyridine ≥98%
H61271.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
:
:
162.02 g/mol
:
GHS05
:
GHS07
:
Ambient
:
58584-94-4
:
C₆H₅Cl₂N
:
MFCD00137847