Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
Supplier:
Cayman Chemical
Certificates
Expand image
D-(-)-Luciferin potassium salt
Supplier:
Cayman Chemical
About this item
CAS Number:
115144-35-9
:
Formula:
C₁₁H₇KN₂O₃S₂
MDL Number:
MFCD00044928
:
318.41 g/mol